Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCR3
Entrez Gene ID
715574
Gene Name
natural cytotoxicity triggering receptor 3
Gene Aliases
NKp30
Chromosome Location
Chr.4: 137919086 - 137923382 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042640.1 NP_001036105.1 - 523 55
XM_015135930.2 XP_014991416.2 - 1051 55
XM_015135931.2 XP_014991417.2 - 907 55
XM_028846835.1 XP_028702668.1 - 800 55
XM_028846836.1 XP_028702669.1 - 656 55
GenBank mRNA AJ554301.1 - - 546 55
AY035214.1 - - 523 55
AY035215.1 - - 523 55
AY035216.1 - - 448 55
AY035217.1 - - 468 55

View Full Product Details