Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APOE
Entrez Gene ID
714623
Gene Name
apolipoprotein E
Gene Aliases
Apo-E
Chromosome Location
Chr.19: 44944814 - 44948739 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028839202.1 XP_028695035.1 - 316 58
GenBank mRNA JU485824.1 - - 320 58
JV667389.1 - - 242 58
U52030.1 - - 28 58

View Full Product Details