Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCC13
Entrez Gene ID
574326
Gene Name
ATP-binding cassette transporter 13
Gene Aliases
-
Chromosome Location
Chr.3: 32490741 - 32582391 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032930.1 NP_001028102.1 - 3103 105
GenBank mRNA AY340665.1 - - 3103 105

View Full Product Details