Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRS3
Entrez Gene ID
574261
Gene Name
bombesin receptor subtype 3
Gene Aliases
EGK_20982
Chromosome Location
Chr.X: 132587123 - 132591388 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032902.1 NP_001028074.1 - 821 79
GenBank mRNA AY350447.1 - - 821 79

View Full Product Details