Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COX7A1
Entrez Gene ID
692060
Gene Name
cytochrome c oxidase subunit 7A1
Gene Aliases
COX7AH
Chromosome Location
Chr.19: 35578016 - 35580114 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040278.2 NP_001035368.1 - 193 62
XM_015123564.2 XP_014979050.1 - 182
GenBank mRNA AY585862.1 - - 146 62
JU323488.1 - - 187 62
JV045060.1 - - 193 62

View Full Product Details