Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP2D6
Entrez Gene ID
678691
Gene Name
cytochrome P450, family 2, subfamily D, polypeptide 6
Gene Aliases
CYP2D17, CYP2D42
Chromosome Location
Chr.10: 9363007 - 9367184 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040218.1 NP_001035308.1 - 842 89
GenBank mRNA AY635464.1 - - 842 89
GU981763.1 - - 850 89
JV708008.1 - - 499 89

Gene Symbol
LOC100424965
Entrez Gene ID
100424965
Gene Name
cytochrome P450 2D17
Gene Aliases
CYP2D44, CYP2D8, CYP2D8P
Chromosome Location
Chr.10: 9346037 - 9351347 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015150493.2 XP_015005979.1 - 941 89

View Full Product Details