Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UNC50
Entrez Gene ID
677700
Gene Name
unc-50 inner nuclear membrane RNA binding protein
Gene Aliases
EGK_05597, GMH1
Chromosome Location
Chr.13: 2378226 - 2393948 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195448.1 NP_001182377.1 - 505 138
XM_015112900.2 XP_014968386.2 - 587 138
GenBank mRNA AY680454.1 - - 189 138
FS723120.1 - - 521 138
JU333890.1 - - 447 138
JV046694.1 - - 428 138
JV046695.1 - - 496 138

View Full Product Details