Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRNP40
Entrez Gene ID
705059
Gene Name
small nuclear ribonucleoprotein U5 subunit 40
Gene Aliases
WDR57
Chromosome Location
Chr.1: 193360165 - 193398998 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195650.1 NP_001182579.1 - 929 97
GenBank mRNA DQ147949.1 - - 337 97
JU477897.1 - - 946 97

View Full Product Details