Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCN2B
Entrez Gene ID
678698
Gene Name
sodium voltage-gated channel beta subunit 2
Gene Aliases
-
Chromosome Location
Chr.14: 111221634 - 111231270 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195475.1 NP_001182404.1 - 83 90
GenBank mRNA DQ190544.1 - - 83 90
JV590291.1 - - 167 90

View Full Product Details