Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFA4
Entrez Gene ID
641333
Gene Name
NDUFA4 mitochondrial complex associated
Gene Aliases
-
Chromosome Location
Chr.3: 104253718 - 104260347 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035524.1 NP_001030601.1 - 121 136
GenBank mRNA DQ148135.1 - - 121 136
JU473786.1 - - 47 136
JV567876.1 - - 179 136

View Full Product Details