Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RMC1
- Entrez Gene ID
-
700967
- Gene Name
- regulator of MON1-CCZ1
- Gene Aliases
-
C18H18orf8, WDR98
- Chromosome Location
-
Chr.18: 58885789 - 58914394 on Build Mmul_10
- Gene Symbol
- NPC1
- Entrez Gene ID
-
700835
- Gene Name
- NPC intracellular cholesterol transporter 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 58831430 - 58886081 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JU334691.1 |
- |
- |
4956 |
91 |
JV565131.1 |
- |
- |
4897 |
91 |
View Full Product Details