Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VTA1
Entrez Gene ID
701578
Gene Name
vesicle trafficking 1
Gene Aliases
-
Chromosome Location
Chr.4: 45896635 - 45975750 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261520.1 NP_001248449.1 - 237 139
XM_028846715.1 XP_028702548.1 - 222 139
GenBank mRNA JU334917.1 - - 231 139
JU475925.1 - - 236 139
JV047321.1 - - 237 139

View Full Product Details