Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APEX1
Entrez Gene ID
702757
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 1
Gene Aliases
EGK_17940
Chromosome Location
Chr.7: 82303407 - 82305972 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258109.1 NP_001245038.1 - 464 60
XM_015143085.2 XP_014998571.1 - 710 60
XM_015143087.2 XP_014998573.1 - 728 60
XM_015143088.2 XP_014998574.1 - 714 60
XM_015143089.2 XP_014998575.1 - 704 60
XM_015143090.2 XP_014998576.1 - 598 60
XM_015143091.2 XP_014998577.1 - 602 60
GenBank mRNA JU326201.1 - - 380 60
JU475401.1 - - 464 60
JV046924.1 - - 700 60
JV046925.1 - - 634 60
JV046926.1 - - 509 60

View Full Product Details