Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX3
Entrez Gene ID
700989
Gene Name
sorting nexin 3
Gene Aliases
EGK_15468
Chromosome Location
Chr.4: 11907568 - 11956849 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257953.1 NP_001244882.1 - 466 104
GenBank mRNA JU320077.1 - - 466 104
JU471826.1 - - 471 104
JV048574.1 - - 498 104
JV597111.1 - - 486 104
JV632129.1 - - 479 104

View Full Product Details