Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STK26
Entrez Gene ID
707739
Gene Name
serine/threonine kinase 26
Gene Aliases
MST4
Chromosome Location
Chr.X: 128272306 - 128325014 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261722.1 NP_001248651.1 - 931 96
XM_015128177.2 XP_014983663.1 - 1263 96
XM_028841795.1 XP_028697628.1 - 1226 96
GenBank mRNA JU472622.1 - - 931 96

View Full Product Details