Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTS
Entrez Gene ID
700189
Gene Name
neurotensin
Gene Aliases
EGK_03983
Chromosome Location
Chr.11: 85501450 - 85508725 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001090068.3 XP_001090068.1 - 540 139

View Full Product Details