Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TACC1
Entrez Gene ID
706243
Gene Name
transforming acidic coiled-coil containing protein 1
Gene Aliases
-
Chromosome Location
Chr.8: 39202846 - 39328788 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JU327040.1 - - 602 121
JU336103.1 - - 2195 121
JU476962.1 - - 2317 121
JU476963.1 - - 1084 121
JV565172.1 - - 2323 121

View Full Product Details