Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HNRNPH3
Entrez Gene ID
694760
Gene Name
heterogeneous nuclear ribonucleoprotein H3
Gene Aliases
EGK_19802
Chromosome Location
Chr.9: 69582142 - 69593110 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266749.1 NP_001253678.1 - 716 75
XM_015147324.2 XP_015002810.1 - 775 75
XM_015147325.2 XP_015002811.1 - 840 75
XM_015147326.2 XP_015002812.1 - 730 75
XM_015147327.2 XP_015002813.1 - 711 75
XM_015147328.2 XP_015002814.1 - 795 75
XM_028853261.1 XP_028709094.1 - 636 75
XM_028853262.1 XP_028709095.1 - 617 75
XM_028853263.1 XP_028709096.1 - 701 75
XM_028853264.1 XP_028709097.1 - 591 75
XM_028853265.1 XP_028709098.1 - 572 75
GenBank mRNA JU320257.1 - - 735 75
JU334281.1 - - 671 75
JU334282.1 - - 716 75
JV649904.1 - - 912 75

View Full Product Details