Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CREG1
Entrez Gene ID
701021
Gene Name
cellular repressor of E1A stimulated genes 1
Gene Aliases
CREG
Chromosome Location
Chr.1: 24904123 - 24917094 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001089325.4 XP_001089325.1 - 567 94
GenBank mRNA JU323139.1 - - 539 94
JU472918.1 - - 539 94
JV703756.1 - - 545 94

View Full Product Details