Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBB
Entrez Gene ID
696110
Gene Name
ubiquitin B
Gene Aliases
-
Chromosome Location
Chr.16: 16007553 - 16009566 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015118664.2 XP_014974150.1 - 415 69

View Full Product Details