Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBFB
Entrez Gene ID
695767
Gene Name
core-binding factor subunit beta
Gene Aliases
-
Chromosome Location
Chr.20: 54207708 - 54284953 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266558.1 NP_001253487.1 - 703 79
XM_028840379.1 XP_028696212.1 - 627 79
GenBank mRNA JU336883.1 - - 580 79
JU472751.1 - - 661 79
JV043621.1 - - 703 79

View Full Product Details