Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEA1
Entrez Gene ID
699300
Gene Name
male-enhanced antigen 1
Gene Aliases
EGK_14921
Chromosome Location
Chr.4: 126325647 - 126328303 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194869.1 NP_001181798.1 - 508 64
XM_001087765.4 XP_001087765.2 - 531 64

Gene Symbol
PPP2R5D
Entrez Gene ID
699163
Gene Name
protein phosphatase 2 regulatory subunit B'delta
Gene Aliases
-
Chromosome Location
Chr.4: 126327350 - 126354769 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JU335298.1 - - 3224 64
JU471102.1 - - 3220 64
JV045007.1 - - 3223 64
JV566389.1 - - 3232 64
JV600899.1 - - 3185 64

View Full Product Details