Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NABP1
Entrez Gene ID
694172
Gene Name
nucleic acid binding protein 1
Gene Aliases
EGK_04635, OBFC2A
Chromosome Location
Chr.12: 78910185 - 78923998 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261728.1 NP_001248657.1 - 544 104
XM_015110573.2 XP_014966059.1 - 658 104
XM_015110575.2 XP_014966061.1 - 459 104
GenBank mRNA JU472958.1 - - 708 104
JU476842.1 - - 570 104
JV044207.1 - - 544 104

View Full Product Details