Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A8
Entrez Gene ID
696345
Gene Name
membrane spanning 4-domains A8
Gene Aliases
EGK_06164, MS4A8B
Chromosome Location
Chr.14: 13499800 - 13514723 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001085445.3 XP_001085445.1 - 222 66
XM_015114062.2 XP_014969548.1 - 467 66
XM_015114063.2 XP_014969549.1 - 468 66

View Full Product Details