Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDP
Entrez Gene ID
702203
Gene Name
norrin cystine knot growth factor NDP
Gene Aliases
EGK_20413
Chromosome Location
Chr.X: 43868809 - 43894161 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266972.1 NP_001253901.1 - 193 77
GenBank mRNA JU320806.1 - - 187 77
JV044452.1 - - 193 77

View Full Product Details