Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFAF4
Entrez Gene ID
705102
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 4
Gene Aliases
C4H6orf66
Chromosome Location
Chr.4: 76403980 - 76412360 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260754.1 NP_001247683.1 - 288 121
GenBank mRNA JU332311.1 - - 321 121
JU475282.1 - - 284 121
JV045453.1 - - 288 121
JV567093.1 - - 317 121
JV638897.1 - - 323 121

View Full Product Details