Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TYW3
Entrez Gene ID
704515
Gene Name
tRNA-yW synthesizing protein 3 homolog
Gene Aliases
-
Chromosome Location
Chr.1: 149638908 - 149673574 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257439.1 NP_001244368.1 - 229 66
XM_015142336.2 XP_014997822.1 - 398 66
XM_015142343.2 XP_014997829.1 - 398 66
GenBank mRNA JU332909.1 - - 229 66
JU474305.1 - - 173 66
JV046184.1 - - 272 66
JV642179.1 - - 219 66

View Full Product Details