Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM23
Entrez Gene ID
712650
Gene Name
RNA binding motif protein 23
Gene Aliases
-
Chromosome Location
Chr.7: 84924813 - 84945155 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266374.1 NP_001253303.1 - 906 59
XM_015143281.2 XP_014998767.2 - 947 59
XM_015143282.2 XP_014998768.2 - 899 59
XM_015143283.2 XP_014998769.2 - 899 59
XM_015143284.2 XP_014998770.2 - 703 59
GenBank mRNA JU335242.1 - - 846 59
JU335243.1 - - 894 59
JU476212.1 - - 874 59
JU476213.1 - - 922 59
JV047508.1 - - 858 59
JV047509.1 - - 906 59
JV527806.1 - - 835 59
JV527807.1 - - 883 59

View Full Product Details