Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM23
Entrez Gene ID
712650
Gene Name
RNA binding motif protein 23
Gene Aliases
-
Chromosome Location
Chr.7: 84924813 - 84945155 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266374.1 NP_001253303.1 - 1040 102
XM_015143281.2 XP_014998767.2 - 1081 102
XM_015143282.2 XP_014998768.2 - 1033 102
XM_015143283.2 XP_014998769.2 - 1033 102
XM_015143284.2 XP_014998770.2 - 837 102
GenBank mRNA JU335242.1 - - 980 102
JU335243.1 - - 1028 102
JU476212.1 - - 1008 102
JU476213.1 - - 1056 102
JV047508.1 - - 992 102
JV047509.1 - - 1040 102
JV527806.1 - - 969 102
JV527807.1 - - 1017 102

View Full Product Details