Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDX1
Entrez Gene ID
706142
Gene Name
peroxiredoxin 1
Gene Aliases
EGK_00700
Chromosome Location
Chr.1: 178900467 - 178920562 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260933.1 NP_001247862.1 - 74 80
GenBank mRNA JV043909.1 - - 74 80

View Full Product Details