Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RHEB
Entrez Gene ID
715378
Gene Name
Ras homolog, mTORC1 binding
Gene Aliases
-
Chromosome Location
Chr.3: 177465325 - 177517200 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261182.1 NP_001248111.1 - 431 93
XM_028845048.1 XP_028700881.1 - 386 93
GenBank mRNA JU320547.1 - - 616 93
JU470465.1 - - 612 93
JV047844.1 - - 431 93

View Full Product Details