Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA1
Entrez Gene ID
701361
Gene Name
serpin family A member 1
Gene Aliases
EGK_18500
Chromosome Location
Chr.7: 156554348 - 156566679 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266017.2 NP_001252946.1 - 656 87
XM_015144331.2 XP_014999817.1 - 1778 87
XM_015144333.2 XP_014999819.1 - 984 87
XM_028850276.1 XP_028706109.1 - 671 87
XM_028850277.1 XP_028706110.1 - 1070 87
GenBank mRNA JU323482.1 - - 657 87
JV704008.1 - - 655 87

View Full Product Details