Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC9
Entrez Gene ID
709303
Gene Name
thioredoxin domain containing 9
Gene Aliases
EGK_05607
Chromosome Location
Chr.13: 3141315 - 3156630 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001104266.4 XP_001104266.1 - 224 113
GenBank mRNA JU332441.1 - - 141 113
JU472973.1 - - 130 113
JV045208.1 - - 102 113

View Full Product Details