Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HS2ST1
Entrez Gene ID
712540
Gene Name
heparan sulfate 2-O-sulfotransferase 1
Gene Aliases
EGK_00936
Chromosome Location
Chr.1: 137403383 - 137598611 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260915.1 NP_001247844.1 - 956 73
GenBank mRNA JU337259.1 - - 661 73
JU477966.1 - - 636 73
JV048806.1 - - 956 73
JV634365.1 - - 976 73

View Full Product Details