Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A20
Entrez Gene ID
709029
Gene Name
solute carrier family 25 member 20
Gene Aliases
EGK_11657
Chromosome Location
Chr.2: 104924007 - 104965475 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265864.1 NP_001252793.1 - 496 78
GenBank mRNA JU321606.1 - - 542 78
JU473369.1 - - 496 78

View Full Product Details