Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BLOC1S2
Entrez Gene ID
709877
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 2
Gene Aliases
EGK_19976
Chromosome Location
Chr.9: 100737992 - 100747761 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266915.1 NP_001253844.1 - 59 63
XM_028853459.1 XP_028709292.1 - 122 63
GenBank mRNA JV600271.1 - - 59 63

View Full Product Details