Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBP2
Entrez Gene ID
715755
Gene Name
retinol binding protein 2
Gene Aliases
EGK_12053
Chromosome Location
Chr.2: 41601689 - 41630121 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194599.2 NP_001181528.1 - 249 99
XM_028843394.1 XP_028699227.1 - 182 99

View Full Product Details