Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANAPC16
Entrez Gene ID
709147
Gene Name
anaphase promoting complex subunit 16
Gene Aliases
EGK_19746
Chromosome Location
Chr.9: 65547906 - 65567929 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257658.1 NP_001244587.1 - 264 116
XM_015147356.2 XP_015002842.1 - 310 116
XM_015147357.2 XP_015002843.1 - 373 116
GenBank mRNA JU332861.1 - - 264 116
JU474264.1 - - 263 116
JV046151.1 - - 203 116

View Full Product Details