Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR2G
Entrez Gene ID
718548
Gene Name
RNA polymerase II subunit G
Gene Aliases
-
Chromosome Location
Chr.14: 11431895 - 11437252 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266524.1 NP_001253453.1 - 117 74
GenBank mRNA JU320820.1 - - 79 74
JU473750.1 - - 52 74
JV047421.1 - - 117 74

View Full Product Details