Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PRKRA
- Entrez Gene ID
-
703063
- Gene Name
- protein activator of interferon induced protein kinase EIF2AK2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 65561754 - 65582527 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JU477604.1 |
- |
- |
1250 |
69 |
- Gene Symbol
- BSG
- Entrez Gene ID
-
721068
- Gene Name
- basigin (Ok blood group)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 331412 - 345288 on Build Mmul_10
View Full Product Details