Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAGLN2
Entrez Gene ID
719527
Gene Name
transgelin 2
Gene Aliases
-
Chromosome Location
Chr.1: 91017933 - 91025320 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265900.1 NP_001252829.1 - 526 79
XM_015110251.2 XP_014965737.1 - 613 79
XM_015110257.2 XP_014965743.1 - 616 79
XM_028846138.1 XP_028701971.1 - 490 79
XM_028846145.1 XP_028701978.1 - 490 79
XM_028846149.1 XP_028701982.1 - 490 79
XM_028846153.1 XP_028701986.1 - 508 79
GenBank mRNA JU336656.1 - - 520 79
JU477453.1 - - 492 79
JV048413.1 - - 526 79
JV663133.1 - - 530 79

View Full Product Details