Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFI27
Entrez Gene ID
700513
Gene Name
interferon alpha inducible protein 27
Gene Aliases
-
Chromosome Location
Chr.7: 156289053 - 156295024 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JU334335.1 - - 390 122
JU475434.1 - - 391 122
JV650231.1 - - 401 122

View Full Product Details