Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC711061
Entrez Gene ID
711061
Gene Name
alcohol dehydrogenase 1C
Gene Aliases
ADH1C
Chromosome Location
Chr.5: 97723507 - 97749859 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001099784.4 XP_001099784.1 - 918 148

View Full Product Details