Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC715623
Entrez Gene ID
715623
Gene Name
aflatoxin B1 aldehyde reductase member 3
Gene Aliases
AKR7A3
Chromosome Location
Chr.1: 205440844 - 205447612 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001105933.4 XP_001105933.3 - 1229 58

View Full Product Details