Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTG1
Entrez Gene ID
710112
Gene Name
BTG anti-proliferation factor 1
Gene Aliases
-
Chromosome Location
Chr.11: 91931999 - 91934664 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266672.1 NP_001253601.1 - 598 162
GenBank mRNA JU320890.1 - - 374 162
JU471743.1 - - 620 162
JU471744.1 - - 620 162
JV044700.1 - - 598 162

View Full Product Details