Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRMT12
Entrez Gene ID
705848
Gene Name
tRNA methyltransferase 12 homolog
Gene Aliases
EGK_19257
Chromosome Location
Chr.8: 124776991 - 124779957 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001101713.4 XP_001101713.4 - 2050 141
GenBank mRNA JU335213.1 - - 1381 141
JU470538.1 - - 1419 141
JV045344.1 - - 1388 141

View Full Product Details