Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FIGN
Entrez Gene ID
702456
Gene Name
fidgetin, microtubule severing factor
Gene Aliases
fidgetin
Chromosome Location
Chr.12: 50725945 - 50860702 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001097992.3 XP_001097992.1 - 496 78

View Full Product Details