Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VRTN
Entrez Gene ID
699513
Gene Name
vertebrae development associated
Gene Aliases
C14orf115, C7H14orf115, EGK_18374
Chromosome Location
Chr.7: 136454434 - 136477657 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028850259.1 XP_028706092.1 - 244 148

View Full Product Details