Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCDHB3
Entrez Gene ID
700243
Gene Name
protocadherin beta 3
Gene Aliases
-
Chromosome Location
Chr.6: 138479861 - 138485835 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001090814.4 XP_001090814.2 - 5230 143

View Full Product Details