Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPR84
Entrez Gene ID
705309
Gene Name
G protein-coupled receptor 84
Gene Aliases
EGK_03734
Chromosome Location
Chr.11: 53977359 - 53979429 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001109742.3 XP_001109742.1 - 122 115

View Full Product Details